Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-4 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB042504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042504 Supplier Novus Biologicals Supplier No. NBP297308AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-4 Polyclonal specifically detects Caspase-4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Caspase-4
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias apoptotic cysteine protease Mih1/TX, CASP-4, caspase 4, apoptosis-related cysteine peptidase, caspase 4, apoptosis-related cysteine protease, caspase-4, EC 3.4.22, EC 3.4.22.57, ICE(rel)II, ICE(rel)-II, ICEREL-II, ICH2, ICH-2, Mih1/TX, Protease ICH-2, Protease TX, TX
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Caspase-4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 837
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.