Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-7 Antibody (25B881.1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10056529C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056529C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056529C Supplier Novus Biologicals Supplier No. NB10056529C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Caspase-7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 25B881.1
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. P55210
Gene Alias apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma
Gene Symbols CASP7
Host Species Mouse
Immunogen This antibody was generated against recombinant protein of active form of human Caspase-7.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 840
Test Specificity In Jurkat, a 35 kDa band is observed.
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.