Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-7 Antibody (7-1-11), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB004224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004224 Supplier Novus Biologicals Supplier No. NBP119229DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat, Porcine, Canine, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Caspase-7
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 7-1-11
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma
Gene Symbols CASP7
Host Species Mouse
Immunogen Recombinant human Caspase-7 protein (full length)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Test Specificity Caspase-7 (7-1-11)
Target Species Human, Mouse, Rat, Pig, Canine, Hamster, Primate, Rabbit, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.