Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-7 Antibody (Mch3 1-1-11), Novus Biologicals™
SDP

Catalog No. NB500206SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500206SS 0.025 mL
NB500206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500206SS Supplier Novus Biologicals Supplier No. NB500206SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockdown Validated.

Specifications

Antigen Caspase-7
Applications Western Blot
Classification Monoclonal
Clone Mch3 1-1-11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Simple Western 1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. P55210
Gene Alias apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma
Gene Symbols CASP7
Host Species Mouse
Immunogen Full-length recombinant human Caspase 7 [UniProt# P55210]
Molecular Weight of Antigen 35 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 840
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.