Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-7 Antibody (Mch3 1-1-11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$207.00 - $501.50

Specifications

Antigen Caspase-7
Clone Mch3 1-1-11
Dilution Western Blot 1:500-1:1000, Simple Western 1:500
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB500206SS
SDP
View Documents
Novus Biologicals
NB500206SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB500206
SDP
View Documents
Novus Biologicals
NB500206
0.1 mL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockdown Validated.
Specifications

Specifications

Caspase-7
Western Blot 1:500-1:1000, Simple Western 1:500
Monoclonal
Ascites
RUO
Human, Mouse, Rat
P55210
840
Full-length recombinant human Caspase 7 [UniProt# P55210]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Mch3 1-1-11
Western Blot
Unconjugated
Mouse
Apoptosis, Cancer, Caspases, Cell Biology, Ubiquitin Proteasome Pathway
Ascites with 0.1% Sodium Azide
apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma
CASP7
IgG1 κ
Unpurified
35 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.