Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-8 Antibody (90A992) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056527T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056527T 0.025 mg
NB10056527 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056527T Supplier Novus Biologicals Supplier No. NB10056527SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

Caspase-8 Monoclonal specifically detects Caspase-8 in Drosophila melanogaster, Human, Primate, Rhesus Macaque samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen Caspase-8
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Monoclonal
Clone 90A992
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Simple Western 1:100, Flow Cytometry 0.1-0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 31034097), Immunohistochemistry-Paraffin 4 ug/ml, CyTOF-ready
Gene Accession No. Q14790
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols CASP8
Host Species Mouse
Immunogen The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Apoptosis Detections, Cancer, Caspases, Necroptosis
Primary or Secondary Primary
Gene ID (Entrez) 841
Test Specificity This can be used for detection of the pro-form of Caspase-8.
Target Species Drosophila melanogaster, Primate, Human, Primate, Rhesus Macaque
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.