Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-8 Antibody (90A992) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
$207.00 - $487.50
Specifications
Antigen | Caspase-8 |
---|---|
Clone | 90A992 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Simple Western 1:100, Flow Cytometry 0.1-0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 31034097), Immunohistochemistry-Paraffin 4 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056527T
![]() |
Novus Biologicals
NB10056527SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056527
![]() |
Novus Biologicals
NB10056527 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Caspase-8 Monoclonal specifically detects Caspase-8 in Human, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Caspase-8 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis, Apoptosis Detections, Cancer, Caspases, Necroptosis | |
Q14790 | |
841 | |
The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
90A992 | |
Western Blot 0.5-2 ug/ml, Simple Western 1:100, Flow Cytometry 0.1-0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 31034097), Immunohistochemistry-Paraffin 4 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
CASP8 | |
IgG1 | |
Protein G purified | |
This can be used for detection of the pro-form of Caspase-8. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title