Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-8 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB193641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB193641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB193641 Supplier Novus Biologicals Supplier No. NBP176610AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-8 Polyclonal antibody specifically detects Caspase-8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Caspase-8
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human Caspase-8 isoform A. The immunogen is located within the last 50 amino acids of Caspase-8. Amino Acid Squence: NYEVSNKDDKKNMGKQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Apoptosis Detections, Cancer, Caspases, Cell Biology, Necroptosis, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 841
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.