Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-8 Antibody - (Pro and Active), Novus Biologicals™
SDP

Catalog No. NBP105123 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105123 0.05 mL
NBP105123SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP105123 Supplier Novus Biologicals Supplier No. NBP105123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Caspase-8 Polyclonal specifically detects Caspase-8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Caspase-8
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. NP_001073595
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols CASP8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human Caspase-8 (PQPTFTLRKKLVFPSD). This sequence is common to various Caspase-8 forms including: 1. Caspase 8a (aa 481-496 GenBank no. NP_001219.2) 2. Caspase 8b (aa 464-479, GenBank no. NP_203519.1) 3. Caspase 8c (aa 449-464, GenBank no. NP_203520.1) 4. Caspase 8L (aa 523-538, GenBank no. gbAAL87628.1) 5. Truncated caspase 8 (as 251-266, GenBank no. gi3928274)
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 841
Test Specificity Caspase-8
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.