Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-8 Antibody - (Pro and Active), Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$214.50 - $330.00

Specifications

Antigen Caspase-8
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP105123SS
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP105123SS
0.025 mL
Each for $214.50
Only null left
Add to Cart
 
NBP105123
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP105123
0.05 mL
Each for $330.00
Only null left
Add to Cart
 
Description

Description

Caspase-8 Polyclonal specifically detects Caspase-8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Caspase-8
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Apoptosis, Caspases
Whole antisera with 0.05% Sodium Azide
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
CASP8
IgG
Unpurified
Caspase-8
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Polyclonal
Antisera
RUO
Human, Mouse, Rat
NP_001073595
841
Synthetic peptide corresponding to the C-terminus of human Caspase-8 (PQPTFTLRKKLVFPSD). This sequence is common to various Caspase-8 forms including: 1. Caspase 8a (aa 481-496 GenBank no. NP_001219.2) 2. Caspase 8b (aa 464-479, GenBank no. NP_203519.1) 3. Caspase 8c (aa 449-464, GenBank no. NP_203520.1) 4. Caspase 8L (aa 523-538, GenBank no. gbAAL87628.1) 5. Truncated caspase 8 (as 251-266, GenBank no. gi3928274)
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.