Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Antibody (LAP6 96-2-22) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500209 0.2 mg
NB500209SS 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500209 Supplier Novus Biologicals Supplier No. NB500209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Caspase-9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LAP6 96-2-22
Concentration 30.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. P55211
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbols CASP9
Host Species Mouse
Immunogen Recombinant human Caspase 9 pro-domain protein. [UniProt# P55211]
Purification Method Unpurified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 842
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.