Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-9 Antibody (LAP6 96-2-22) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | Caspase-9 |
---|---|
Clone | LAP6 96-2-22 |
Concentration | 30.7 mg/mL |
Dilution | Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500209SS
![]() |
Novus Biologicals
NB500209SS |
0.05 mg |
Each for $206.50
|
|
|||||
NB500209
![]() |
Novus Biologicals
NB500209 |
0.2 mg |
Each for $499.50
|
|
|||||
Description
Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Caspase-9 | |
30.7 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Caspases, Cell Biology | |
Ascites with 0.1% Sodium Azide | |
APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
CASP9 | |
IgG1 κ | |
Unpurified |
LAP6 96-2-22 | |
Western Blot 1 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 8-25 ug/ml. Use reported by customer review, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 8-25 ug/ml, KnockDown Validated | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
P55211 | |
842 | |
Recombinant human Caspase 9 pro-domain protein. [UniProt# P55211] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title