Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356950 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356950 Supplier Novus Biologicals Supplier No. NBP176961MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-9 Polyclonal antibody specifically detects Caspase-9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunofluorescence, Immunocytochemistry

Specifications

Antigen Caspase-9
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 20 amino acids near the center of human Caspase 9. The immunogen is located within amino acids 290-340 of Caspase 9. Amino Acid Sequence: ASTSPEDESPGSNPEPDATP
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 842
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.