Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Inhibitor III, Thermo Scientific Chemicals

Catalog No. aaj65501mcr
Click to view available options
Quantity:
1 mg

A potent and irreversible inhibitor of caspase-9

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403848-57-7
Molecular Formula C24H35ClN6O9
Synonym Ac-LEHD-CMK;

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A potent and irreversible inhibitor of caspase-9
Formula Weight 587.02
Form Powder
InChI Key JSRLQUNMFYXVQD-UHFFFAOYNA-N
SMILES CC(C)CC(NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CN=CN1)C(=O)NC(CC(O)=O)C(=O)CCl
IUPAC Name 3-{2-[4-carboxy-2-(2-acetamido-4-methylpentanamido)butanamido]-3-(1H-imidazol-5-yl)propanamido}-5-chloro-4-oxopentanoic acid
Molecular Weight (g/mol) 587.03
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.