Learn More
Promega Caspase-Glo™ 3/7 Assay Kit
Description
Includes
Caspase-Glo 3/7 Substrate; Caspase-Glo 3/7 Buffer
Utilizes a proluminescent Caspase-3/7 DEVD-aminoluciferin substrate and a proprietary thermostable luciferase in a single reagent optimized for Caspase-3/7 activity, luciferase activity, and cell lysis. Stabilized luciferase improves assay performance across a wide range of assay conditions and makes the assay less likely to be affected by compound interference than other luminescent-, fluorescent-, or colorimetric-based assays.
- Designed for use with 96- or 384-well plates using either purified enzyme or cells in culture as sample
- Requires less enzyme or fewer cells than other caspase formats due to low background luminescence
- Requires no sample preparation, manipulation, or extended incubation times—maximum sensitivity is achieved in as little as one-half to one hour
- “Add-mix-measure” protocol makes the assay easy to automate
- Extended-glow signal allows the plates to be read over a three-hour period for batch processing—no injectors required
- Patent pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patent Nos. 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
Quantity | 10 x 100 mL |
pH | 7.4 |
Content And Storage | -20°C |
For Use With (Application) | 96- or 384-well plates using either purified enzyme or cells in culture as sample |
Form | Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.