Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Caspase-Glo™ 6 Assay System
Click to view available options
Quantity:
10 mL
50 mL
Description
- Advantages: simplified method, greater sensitivity provide faster results, increased accuracy
- Batch-processing capability
- Includes: Caspase-Glo 6 Buffer, Z-VEID-Glo™ Substrate, Luciferin Detection Reagent
- Ideal for biochemical screens for Caspase -2 inhibitors
Specifications
Specifications
Quantity | 50 mL |
pH | 7.2 |
Kit Contents | 1 x 50mL Caspase-Glo 6 Buffer, 1 x 500μL Z-VEID-Glo substrate, 1 x 1 each Luciferin Detection Reagent |
Test Time | 30 min. |
Content And Storage | -20°C |
For Use With (Application) | Caspase-6 Enzyme Studies, Caspase-6 Inhibitor Screening |
Form | Fluid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction