Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspr2 Rabbit anti-Human, Mouse, Rat, Clone: 2H2I5, Novus Biologicals™
SDP

Catalog No. NB166542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166542 20 μg
NB166541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166542 Supplier Novus Biologicals Supplier No. NBP31572320UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Caspr2 Monoclonal antibody specifically detects Caspr2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Caspr2
Applications Western Blot
Classification Monoclonal
Clone 2H2I5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1232-1331 of human Caspr2 (Q9UHC6). HLDHLDSASADFPYNPGQGQAIRNGVNRNSAIIgGVIAVVIFTILCTLVFLIRYMFRHKGTYHTNEAKGAESAESADAAIMNNDPNFTETIDESKKEWLI
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 26047
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.