Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASZ1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177942 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177942 Supplier Novus Biologicals Supplier No. NBP177942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASZ1 Polyclonal specifically detects CASZ1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CASZ1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.3 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000-1:50000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. B3KRV8
Gene Alias castor, castor homolog 1, zinc finger, castor homolog 1, zinc finger (Drosophila), castor zinc finger 1, Castor-related protein, CST, dJ734G22.1, FLJ20321, Putative survival-related protein, SRG, SRGcst, survival related, survival-related, Zinc finger protein 693, zinc finger protein castor homolog 1, ZNF693, ZNF693FLJ12223
Gene Symbols CASZ1
Host Species Rabbit
Immunogen This affinity purified CASZ1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to region near the amino terminal end of human CASZ1 protein. (Uniprot: B3KRV8)
Molecular Weight of Antigen 124.82 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54897
Test Specificity Expect cross reactivity with multiple isoforms of this protein.
Target Species Human, Mouse, Rat, Bovine, Primate
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.