Learn More
CASZ1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | CASZ1 |
---|---|
Concentration | 1.07 mg/mL |
Applications | ELISA, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2004
![]() ![]() |
Rockland Immunochemicals
600401B62 |
100 μg |
Each for $636.24
|
|
|||||
50-199-2005
![]() ![]() |
Rockland Immunochemicals
600401B62S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human CASZ1 protein. A BLAST analysis was used to suggest reactivity with CASZ1 proteins from human, mouse, Drosophila, chimpanzee, and macaque based on a 100% homology. Partial reactivity is expected with horse and dog CASZ1 based on a 92% homology with the immunizing sequence. Cross-reactivity with CASZ1 from other sources has not been determined.
The protein encoded by this gene is a zinc finger transcription factor. The encoded protein may function as a tumor suppressor, and single nucleotide polymorphisms in this gene are associated with blood pressure variation. Alternative splicing results in multiple transcript variants that encode different protein isoforms.Specifications
CASZ1 | |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Chimpanzee, Drosophila, Human, Mouse, Rhesus Monkey | |
Q86V15, Q9CWL2 | |
457931, 54897, 69743, 722677 | |
Primary | |
Antibody |
1.07 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
2410019P08Rik; AV096627; BC040081; BOS_15820; CAS11; castor homolog 1, zinc finger; castor zinc finger 1; Castor-related protein; Casz1; Cst; D4Ertd432e; dJ734G22.1; Kiaa3026; Putative survival-related protein; RGD1563533; sb:eu580; SRG; survival-related; wu:fk85f10; Zinc finger protein 693; Zinc finger protein castor homolog 1; ZNF693 | |
CASZ1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
CASZ1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.