Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAT3/SLC7A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174144
Description
CAT3/SLC7A3 Polyclonal specifically detects CAT3/SLC7A3 in Mouse samples. It is validated for Western Blot.Specifications
CAT3/SLC7A3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ATRC3cationic amino acid transporter 3, CAT3, CAT-3Cationic amino acid transporter y+, FLJ14541, MGC20687, solute carrier family 7 (cationic amino acid transporter, y+ system), member 3, Solute carrier family 7 member 3 | |
Rabbit | |
Affinity purified | |
RUO | |
84889 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
P70423 | |
SLC7A3 | |
Synthetic peptides corresponding to the middle region of Slc7a3. Immunizing peptide sequence RAWSSAFDNLIGNHISRTLKGTILLKMPHVLAEYPDFFALALVLLLTGLL. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 76%; Chicken: 76%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction