Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB090261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090261 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090261 Supplier Novus Biologicals Supplier No. NBP299933UV

Rabbit Polyclonal Antibody

Catalase Polyclonal specifically detects Catalase in Human samples. It is validated for ELISA.

Specifications

Antigen Catalase
Applications ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias catalase, EC 1.11.1.6, MGC138422, MGC138424
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Catalase (P04040; Ala2-Leu527).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 847
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.