Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase Antibody (HPYL/7171), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB179278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179278 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179278 Supplier Novus Biologicals Supplier No. NBP314082C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Catalase Monoclonal antibody specifically detects Catalase in H. Pylori samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Catalase
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HPYL/7171
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias catalase, EC 1.11.1.6, MGC138422, MGC138424
Host Species Mouse
Immunogen Recombinant Helicobacter pylori Catalase protein fragment (around aa323-445) (Exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 847
Target Species H. pylori
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.