Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase Antibody (OTI1B8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N270342A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270342A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270342A700 Supplier Novus Biologicals Supplier No. NBP270342AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Catalase Monoclonal specifically detects Catalase in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Catalase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B8
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias catalase, EC 1.11.1.6, MGC138422, MGC138424
Gene Symbols CAT
Host Species Mouse
Immunogen Full length recombinant protein of human Catalase (NP_111743) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 847.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.