Learn More
MP Biomedicals™ Catalase from Human Erythrocytes (E.C.1.11.1.6.)
Description
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).
Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.
Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).

Specifications
Specifications
Concentration | 1 mg/mL |
For Use With (Application) | Protecting cells against oxidative damage to proteins, lipids and nucleic acids |
Storage Buffer | Frozen solution of 1mg/mL in 1.5mM Sodium Phosphate, 20mM NaCl, pH 7.0 |
Content And Storage | -20°C |
Source | Human erythrocyte |
Quantity | 100 kU |
Product Type | Catalase |
Molecular Weight (g/mol) | 256000 |
Form | Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.