Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalase, Lyophilized, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 g
10 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Catalase, Lyophilized , a tetramer of four polypeptide chains, is used in the food industry to remove hydrogen peroxide from milk in order to make cheese. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| CAS | 9001-05-2 |
| Quantity | 5 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction