Learn More
Catalase Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Catalase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0786
![]() ![]() |
Rockland Immunochemicals
200401051 |
100 μg |
Each for $636.24
|
|
|||||
50-199-0787
![]() ![]() |
Rockland Immunochemicals
200401051S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Catalase Antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Catalase [Bovine liver]. Cross reactivity against Catalase from other tissues and species may occur but have not been specifically determined.
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.Specifications
Catalase | |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Bovine, Human, Mouse, Ovine, Rat | |
P00432, P04040, P04762, P24270 | |
100307035, 12359, 24248, 531682, 847 | |
Anti-Catalase Antibody was produced by repeated immunizations with bovine liver catalase protein. | |
-20°C, Avoid Freeze/Thaw Cycles | |
Cat |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
2210418N07; BOS_15095; Cas1; Cas-1; CAT; Cat01; Catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; MGC138422; MGC138424 | |
Cat | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.