Learn More
Catechol, 99%
Description
Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.
Solubility
Soluble in water, pyridine, chloroform, benzene, carbon tetrachloride, ether and acetate.
Notes
Air and light sensitive. Incompatible with oxidizing agents, aluminum, bases, acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | 104°C to 107°C |
| Density | 1.370 |
| Boiling Point | 245°C to 246°C |
| Flash Point | 127°C (260°F) |
| Assay Percent Range | 99% |
| Quantity | 250 g |
| UN Number | UN2811 |
| Beilstein | 471401 |
| Sensitivity | Air and light sensitive |
| Merck Index | 14,7999 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311-H315-H317-H318-H332-H341-H350-H500 |
| Hazard Statement | GHS H Statement H301-H311-H318-H341-H351-H315 |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P333+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 204-427-5 |
| RTECSNumber | UX1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.