Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin alpha 1 Antibody (1G5), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB008338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008338 Supplier Novus Biologicals Supplier No. NBP254533DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Catenin alpha 1 Monoclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Catenin alpha 1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1G5
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13
Gene Symbols CTNNA1
Host Species Mouse
Immunogen Human alpha catenin fusion protein (Uniprot: P35221)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Core ESC Like Genes, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.