Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catenin delta-2 Mouse anti-Human, Mouse, Unconjugated, Clone: 4H2.1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | Catenin delta-2 |
---|---|
Clone | 4H2.1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN2330-25
|
MilliporeSigma
MABN233025UG |
25 μg |
Each for $192.35
|
|
|||||
MABN2330-100
|
MilliporeSigma
MABN2330100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-Catenin delta-2, clone 4H2.1 Antibody, Cat. No. MABN2330, is a highly specific mouse monoclonal antibody that targets Catenin delta-2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
Catenin delta-2 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Delta-catenin;GT24;Neural plakophilin-related ARM-repeat protein;NPRAP;Neurojungin | |
CTNND2;NPRAP | |
IgG2b κ | |
Protein G purified | |
Clone 4H2.1 specifically detects Catenin delta-2 in human and murine cells. It targets an epitope with in the C-tereminal region. | |
∽150 kDa observed; 132.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
4H2.1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Neuroscience | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001323 | |
GST/His-tagged recombinant fragment corresponding to 145 amino acids from the C-terminal region of human Catenin delta-2. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title