Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin 3 Antibody (RM0156-2K13), PE, Novus Biologicals™
SDP

Catalog No. NB360928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360928 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360928 Supplier Novus Biologicals Supplier No. NBP212422PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cathepsin 3 Monoclonal antibody specifically detects Cathepsin 3 in Mouse samples. It is validated for Western Blot

Specifications

Antigen Cathepsin 3
Applications Western Blot
Classification Monoclonal
Clone RM0156-2K13
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias 1600000I23Rik
Host Species Rat
Immunogen Mouse recombinant Cathepsin 3
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117066
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.