Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin 7 Antibody (RM0158-4A12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212424UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212424UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212424UV Supplier Novus Biologicals Supplier No. NBP212424UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cathepsin 7 Monoclonal specifically detects Cathepsin 7 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Cathepsin 7
Applications Western Blot
Classification Monoclonal
Clone RM0158-4A12
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CTS1, Epcs24, Epcs71
Gene Symbols Cts7
Host Species Rat
Immunogen Mouse recombinant Cathepsin 7
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 56092
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.