Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin 7 Antibody (RM0158-4A12), FITC, Novus Biologicals™
SDP

Catalog No. NB360919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360919 Supplier Novus Biologicals Supplier No. NBP212424F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cathepsin 7 Monoclonal antibody specifically detects Cathepsin 7 in Mouse samples. It is validated for Western Blot

Specifications

Antigen Cathepsin 7
Applications Western Blot
Classification Monoclonal
Clone RM0158-4A12
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias CTS1, Epcs24, Epcs71
Host Species Rat
Immunogen Mouse recombinant Cathepsin 7
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56092
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.