Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin A/Lysosomal Carboxypeptidase A Antibody (104), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB108160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108160 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108160 Supplier Novus Biologicals Supplier No. NBP289526V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cathepsin A/Lysosomal Carboxypeptidase A
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 104
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin A/Lysosomal Carboxypeptidase A (NP_001121167.1; Met1-Tyr480).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5476
Test Specificity No cross-reactivity in ELISA with: Mouse CTSA Human CTSZ
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.