Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin A/Lysosomal Carboxypeptidase A Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB321198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321198 Supplier Novus Biologicals Supplier No. NBP298264PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cathepsin A/Lysosomal Carboxypeptidase A Polyclonal antibody specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Cathepsin A/Lysosomal Carboxypeptidase A
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Cathepsin A/Lysosomal Carboxypeptidase A (Uniprot#: P16675-1; Met1-Tyr474)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5476
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.