Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin A/Lysosomal Carboxypeptidase A Antibody (RM0160-4A12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP212071B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212071B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212071B Supplier Novus Biologicals Supplier No. NBP212071B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Cathepsin A/Lysosomal Carboxypeptidase A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0160-4A12
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase
Gene Symbols CTSA
Host Species Rat
Immunogen Mouse recombinant Cathepsin A/Lysosomal Carboxypeptidase A
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5476
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.