Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin B Antibody (010), PE, Novus Biologicals™
SDP

Catalog No. NB333336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333336 Supplier Novus Biologicals Supplier No. NBP290402PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cathepsin B Monoclonal antibody specifically detects Cathepsin B in Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cathepsin B
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 010
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, cysteine protease, EC 3.4.22, EC 3.4.22.1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Cathepsin B (Uniprot#: P10605, Met1-Phe339)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1508
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.