Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin D Antibody (003), DyLight 350
SDP

Catalog No. NB037501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037501 Supplier Novus Biologicals Supplier No. NBP290117UV

Rabbit Monoclonal Antibody

Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cathepsin D
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 3
Conjugate DyLight 350
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin D (P07339; Met1-Leu412).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1509
Test Specificity Has cross-reactivity in ELISA with: Mouse CTSD No cross-reactivity in ELISA with: Human CTSA Human CTSB Human CTSL Human BACE1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.