Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin D Antibody (102), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB333233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333233 Supplier Novus Biologicals Supplier No. NBP290432MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cathepsin D Monoclonal antibody specifically detects Cathepsin D in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Cathepsin D
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 102
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Cathepsin D (Accession#: NP_034113.1, Met 1-Leu 410)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1509
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.