Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin D Mouse anti-Human, Clone: 2F6F7, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 665341IG150UL

Description
This antibody is specific to CTSD.
Cathepsin D has been reported to be involved in maligt tumor progression and with prognosis of various human cancers.Specifications
Cathepsin D | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07339 | |
CTSD | |
Cathepsin D Fusion Protein Ag15254 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
2F6F7 | |
Unconjugated | |
CTSD | |
cathepsin D, CLN10, CPSD, CTSD | |
Mouse | |
Protein A | |
RUO | |
1509 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction