Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin D Mouse anti-Human, Clone: 2F6F7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Cathepsin D |
---|---|
Clone | 2F6F7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6955
![]() ![]() |
Proteintech Group Inc
665341IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to CTSD.
Cathepsin D has been reported to be involved in maligt tumor progression and with prognosis of various human cancers.Specifications
Cathepsin D | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
cathepsin D, CLN10, CPSD, CTSD | |
CTSD | |
IgG2b | |
Protein A | |
Antibody |
2F6F7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P07339 | |
1509 | |
Cathepsin D Fusion Protein Ag15254 | |
Primary | |
-20°C | |
CTSD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title