Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin D Rabbit anti-Human, Clone: 3, Novus Biologicals™
SDP

Catalog No. NB021697 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021697 100 μg
NB021698 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021697 Supplier Novus Biologicals Supplier No. NBP290117100UL

Rabbit Monoclonal Antibody

Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cathepsin D
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 3
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Host Species Rabbit
Immunogen Recombinant Human Cathepsin D protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1509
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.