Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin E Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB081839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081839 Supplier Novus Biologicals Supplier No. NBP298303H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cathepsin E Polyclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA.

Specifications

Antigen Cathepsin E
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cathepsin E (NP_031825.2; Met 1-Pro 397).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1510
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.