Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin K Antibody (CTSK/2791), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB011900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011900 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011900 Supplier Novus Biologicals Supplier No. NBP279889AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cathepsin K
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CTSK/2791
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD
Gene Symbols CTSK
Host Species Mouse
Immunogen Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) (Uniprot: P43235)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.