Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin K Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Cathepsin K |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543914
|
Invitrogen™
MA543914 |
20 μL |
Each for $213.50
|
|
|||||
PIMA543913
|
Invitrogen™
MA543913 |
100 μL |
Each for $522.50
|
|
|||||
Description
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.Specifications
Cathepsin K | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
AI323530; Cat K; cathepsin K; Cathepsin O; cathepsin O1; cathepsin O2; Cathepsin X; catK; CTS02; CTSK; CTSO; CTSO1; CTSO2; MGC23107; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; PKND; PYCD | |
CTSK | |
IgG2b κ | |
Protein A/G | |
Antibody |
C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Pig | |
O35186, P43235, P55097, Q9GLE3 | |
13038, 1513, 29175, 397569 | |
Ala115-Met329 (Accession P43235), with N-terminal His Tag. | |
Primary | |
-20°C | |
CTSK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title