Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin S Antibody (054), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB076807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076807 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076807 Supplier Novus Biologicals Supplier No. NBP290713C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cathepsin S Monoclonal specifically detects Cathepsin S in Mouse samples. It is validated for ELISA.

Specifications

Antigen Cathepsin S
Applications ELISA
Classification Monoclonal
Clone 54
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin S (AAB94925.1; Met1-Ile340).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1520
Test Specificity No cross-reactivity in ELISA with: Human CTSS Mouse CTSA Mouse CTSB Mouse CTSC Mouse CTSD Mouse CTSE Mouse CTSX
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.