Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin S Antibody (1F9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP242634G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242634G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242634G Supplier Novus Biologicals Supplier No. NBP242634G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cathepsin S Monoclonal specifically detects Cathepsin S in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cathepsin S
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1F9
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Gene Symbols CTSS
Host Species Mouse
Immunogen Recombinant human CTSS (17-331aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1520
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.