Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin V Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB293318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293318 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293318 Supplier Novus Biologicals Supplier No. NBP326082UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cathepsin V Polyclonal antibody specifically detects Cathepsin V in Human samples. It is validated for ELISA

Specifications

Antigen Cathepsin V
Applications ELISA
Classification Polyclonal
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSUcathepsin L2, preproprotein, CTSVMGC125957, EC 3.4.22, EC 3.4.22.43
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human Cathepsin V (Accession#: NP_001324.2, Met1-Val334)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1515
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.