Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin X/Z/P Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Cathepsin Z |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cathepsin X/Z/P Polyclonal specifically detects Cathepsin X/Z/P in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cathepsin Z | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q9UBR2 | |
1522 | |
This antibody was developed against a recombinant protein corresponding to amino acids: VDGVNYASITRNQHIPQYCGSCWAHASTSAMADRINIKRKGAWPSTLLSVQNVIDCGNAGSCEGGNDLSVW | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Epitope Tags | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
CTSZ | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title