Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CATSPERE Antibody, Novus Biologicals™
SDP

Catalog No. NBP23784920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23784920 20 μL
NBP237849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23784920 Supplier Novus Biologicals Supplier No. NBP23784920UL

Rabbit Polyclonal Antibody

CATSPERE Polyclonal specifically detects CATSPERE in Human samples. It is validated for Western Blot.

Specifications

Antigen C1orf101
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q5SY80.2
Gene Symbols C1orf101
Host Species Rabbit
Immunogen The immunogen for Anti-CA101 antibody is: synthetic peptide directed towards the N-terminal region of Human CA101.
Molecular Weight of Antigen 91 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257044
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.