Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cav1.4 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018495 0.1 mg
NBP130667 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018495 Supplier Novus Biologicals Supplier No. NBP1306670.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cav1.4 Polyclonal specifically detects Cav1.4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Cav1.4
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Sterile water with No Preservative
Gene Alias AIED, CACNAF1, calcium channel, voltage-dependent, L type, alpha 1F subunit, Cav1.4, Cav1.4alpha1, COD4, CORDX, CORDX3CSNB2, CSNB2AAland island eye disease (Forsius-Eriksson ocular albinism, ocular albinismtype 2), CSNBX2, JM8, JMC8, OA2COD3, voltage-dependent L-type calcium channel subunit alpha-1F, Voltage-gated calcium channel subunit alpha Cav1.4
Gene Symbols CACNA1F
Host Species Rabbit
Immunogen A synthetic peptide from a cytoplasmic domain of human CACNA1F (Cav1.4) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 778
Test Specificity Appears to be specific for CACNA1F.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.